Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl Ether, puriss., ≥99.5% (GC), Honeywell™ Riedel-de Haën™

Click to view available options
Quantity:
130g
1L
2.5L
5L
6 x 130g
4 x 2.5L
4 x 5L
6 x 1L
135kg
240L
Packaging:
Aluminum Bottle
Glass Bottle
Metal Drum
60-29-7
C4H10O
74.123
MFCD00011646
RTZKZFJDLAIYFH-UHFFFAOYSA-N
diethyl ether,ether,ethyl ether,diethyl oxide,ethyl oxide,aether,pronarcol,anesthetic ether,3-oxapentane,anaesthetic ether
3283
CHEBI:35702
ethoxyethane
CCOCC
60-29-7
C4H10O
74.123
MFCD00011646
RTZKZFJDLAIYFH-UHFFFAOYSA-N
diethyl ether,ether,ethyl ether,diethyl oxide,ethyl oxide,aether,pronarcol,anesthetic ether,3-oxapentane,anaesthetic ether
3283
CHEBI:35702
ethoxyethane
CCOCC

Puriss., meets analytical specification of BP, Ph. Eur.,≥99.5% (GC)

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
Linear Formula (CH3CH2)2O
UN Number UN1155
Beilstein 1696894
Formula Weight 74.12g/mol
Percent Purity ≥99.5% (GC)
Grade puriss.
Density 0.706g/mL at 25°C
Explosion Limit 0.365
Ignition Point 320°F
Packaging Glass Bottle
Vapor Density 2.6 (vs air)
Flash Point −45°C (−49°F)
Physical State Liquid
Boiling Point 34.6°C
Melting Point -116°C
Quantity 130g
Chemical Name or Material Diethyl Ether, Meets analytical specification of Ph.Eur.BP
Show More Show Less

P210-P261

H224-H302-H336

DOTInformation : class 3 - PG 1 - Diethyl ether

EINECSNumber : 200-467-2

RTECSNumber : KI5775000