Alfa Aesar™ Diethyl ethylmalonate, 99%

Manufacturer: Alfa Aesar™ B2407722

Quantity 100g

 View more versions of this product

Catalog No. AAB2407722

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 133-13-1
    Molecular Formula C9H16O4
    Formula Weight 188.22
    MDL Number MFCD00009167
    Beilstein 607635
    Synonym Ethylmalonic acid diethyl ester; Ethylpropanedioic acid diethyl ester
    Chemical Name or Material Diethyl ethylmalonate

    Specifications

    Formula Weight 188.22
    TSCA TSCA
    MDL Number MFCD00009167
    Health Hazard 2 GHS H Statement
    H227
    Combustible liquid.
    Health Hazard 3 GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Flash Point 88°C (190°F)
    Boiling Point 204° to 206°C
    Melting Point 0°C
    Molecular Formula C9H16O4
    CAS 133-13-1
    Beilstein 607635
    Synonym Ethylmalonic acid diethyl ester; Ethylpropanedioic acid diethyl ester
    Health Hazard 1 Warning
    Quantity 100g
    Chemical Name or Material Diethyl ethylmalonate
    Assay 99%

    GHS H Statement
    H227
    Combustible liquid.

    GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit