Alfa Aesar™ Diethyl ethylphosphonate, 98%

Manufacturer: Alfa Aesar™ A110460B

Quantity 1000g

 View more versions of this product

Catalog No. AAA110460B

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 78-38-6
    Molecular Formula C6H15O3P
    Formula Weight 166.16
    MDL Number MFCD00039888
    UN Number UN3082
    Beilstein 1756861
    Synonym Diethyl ethanephosphonate; Ethylphosphonic acid diethyl ester
    Chemical Name or Material Diethyl ethylphosphonate

    Specifications

    Formula Weight 166.16
    Health Hazard 3 GHS P Statement
    P280-P264-P305+P351+P338-P301+P312-P337+P313-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash thoroughly after handling.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    If eye irritation persists: Get medical advice/attention.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Flash Point 105°C (221°F)
    Synonym Diethyl ethanephosphonate; Ethylphosphonic acid diethyl ester
    Health Hazard 1 Warning
    TSCA TSCA
    Molecular Formula C6H15O3P
    MDL Number MFCD00039888
    UN Number UN3082
    Boiling Point 82° to 83°C (11mmHg)
    Health Hazard 2 GHS H Statement
    H302-H319
    Harmful if swallowed.
    Causes serious eye irritation.
    RTECS Number SZ7925000
    CAS 78-38-6
    Beilstein 1756861
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
    Quantity 1000g
    Chemical Name or Material Diethyl ethylphosphonate
    Assay 98%

    Description

    Applications
    Diethyl ethylphosphonate is used for heavy-metal extraction and solvent separation. It is also used as a gasoline additive, antifoam agent, plasticizer and textile conditioner. Further, it serves as an antistatic agent, flame retardant and as a general reagent in phosphinoalkene and phosphinoalkyne synthesis.

    Solubility
    Miscible with alcohol, ether and organic solvents. Slightly miscible with water.

    Notes
    Incompatible with strong oxidizing agents.

    GHS H Statement
    H302-H319
    Harmful if swallowed.
    Causes serious eye irritation.

    GHS P Statement
    P280-P264-P305+P351+P338-P301+P312-P337+P313-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash thoroughly after handling.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    If eye irritation persists: Get medical advice/attention.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit