Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl ethylphosphonate, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
78-38-6
C6H15O3P
166.157
MFCD00039888
AATNZNJRDOVKDD-UHFFFAOYSA-N
diethyl ethylphosphonate,phosphonic acid, ethyl-, diethyl ester,diethyl ethanephosphonate,ethanephosphonic acid, diethyl ester,unii-y52q01p44t,ccris 6230,diethylethanephosphonate,ethylphosphonic acid diethyl ester,phosphonic acid, p-ethyl-, diethyl ester,diethylethylphosphonate
6534
1-[ethoxy(ethyl)phosphoryl]oxyethane
CCOP(=O)(CC)OCC
78-38-6
C6H15O3P
166.157
MFCD00039888
AATNZNJRDOVKDD-UHFFFAOYSA-N
diethyl ethylphosphonate,phosphonic acid, ethyl-, diethyl ester,diethyl ethanephosphonate,ethanephosphonic acid, diethyl ester,unii-y52q01p44t,ccris 6230,diethylethanephosphonate,ethylphosphonic acid diethyl ester,phosphonic acid, p-ethyl-, diethyl ester,diethylethylphosphonate
6534
1-[ethoxy(ethyl)phosphoryl]oxyethane
CCOP(=O)(CC)OCC
for C-C Bond Formation (Olefination)
Quantity 5g
Packaging Glass bottle
Refractive Index 1.4155 to 1.4175 (20°C, 589nm)
Infrared Spectrum Authentic
Color Colorless
Assay Percent Range 98%
Formula Weight 166.16
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material Diethyl ethylphosphonate

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 201-111-9

RTECSNumber : SZ7925000

TSCA : TSCA