Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Diethyl hexafluoroglutarate, 97%

Manufacturer:  Alfa Aesar™ B2242803

Quantity 1g

 View more versions of this product

Catalog No. AAB2242803


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

424-40-8
296.16
1804802
Diethyl hexafluoroglutarate
C9H10F6O4
MFCD00015156
Diethyl perfluoroglutarate; Hexafluoroglutaric acid diethyl ester

Specifications

224°C
424-40-8
296.16
1804802
Diethyl hexafluoroglutarate
1g
C9H10F6O4
MFCD00015156
Diethyl perfluoroglutarate; Hexafluoroglutaric acid diethyl ester
97%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H332-H315-H319-H335-H227
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

RTECSNumber : MA3750500

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit