Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Diethyl hexafluoroglutarate, 97%

Click to view available options
Quantity:
1g
5g
25g
424-40-8
C9H10F6O4
296.165
MFCD00015156
MSDPXVBLFJODJO-UHFFFAOYSA-N
diethyl hexafluoroglutarate,diethyl perfluoroglutarate,diethyl hexafluoropentanedioate,hexafluoroglutaric acid diethyl ester,pentanedioic acid, hexafluoro-, diethyl ester,glutaric acid, hexafluoro-, diethyl ester,hexafluoropentanedioic acid diethyl ester,diethylester kyseliny hexafluorglutarove,diethylester kyseliny hexafluorglutarove czech,pentanedioic acid, 2,2,3,3,4,4-hexafluoro-, 1,5-diethyl ester
9874
diethyl 2,2,3,3,4,4-hexafluoropentanedioate
CCOC(=O)C(C(C(C(=O)OCC)(F)F)(F)F)(F)F
424-40-8
C9H10F6O4
296.165
MFCD00015156
MSDPXVBLFJODJO-UHFFFAOYSA-N
diethyl hexafluoroglutarate,diethyl perfluoroglutarate,diethyl hexafluoropentanedioate,hexafluoroglutaric acid diethyl ester,pentanedioic acid, hexafluoro-, diethyl ester,glutaric acid, hexafluoro-, diethyl ester,hexafluoropentanedioic acid diethyl ester,diethylester kyseliny hexafluorglutarove,diethylester kyseliny hexafluorglutarove czech,pentanedioic acid, 2,2,3,3,4,4-hexafluoro-, 1,5-diethyl ester
9874
diethyl 2,2,3,3,4,4-hexafluoropentanedioate
CCOC(=O)C(C(C(C(=O)OCC)(F)F)(F)F)(F)F
Quantity 25g
Boiling Point 224°C
Beilstein 1804802
Formula Weight 296.16
Percent Purity 97%
Chemical Name or Material Diethyl hexafluoroglutarate

GHS H Statement
H332-H315-H319-H335-H227
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

RTECSNumber : MA3750500

TSCA : TSCA