Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl Isopropyl Malonate 98%, Acros Organics

Click to view available options
Quantity:
25mL
759-36-4
C10H18O4
202.25
MFCD00040491
BYQFBFWERHXONI-UHFFFAOYSA-N
diethyl isopropylmalonate,diethyl 2-isopropylmalonate,isopropyl diethyl malonate,1,3-diethyl 2-propan-2-yl propanedioate,diethylisopropylmalonate,isopropylmalonic acid diethyl ester,propanedioic acid, 1-methylethyl-, diethyl ester,ethyl isopropylmalonate,1,3-diethyl 2-isopropylpropanedioate,2-isopropyl-malonic acid diethyl ester
12966
diethyl 2-propan-2-ylpropanedioate
CCOC(=O)C(C(C)C)C(=O)OCC
759-36-4
C10H18O4
202.25
MFCD00040491
BYQFBFWERHXONI-UHFFFAOYSA-N
diethyl isopropylmalonate,diethyl 2-isopropylmalonate,isopropyl diethyl malonate,1,3-diethyl 2-propan-2-yl propanedioate,diethylisopropylmalonate,isopropylmalonic acid diethyl ester,propanedioic acid, 1-methylethyl-, diethyl ester,ethyl isopropylmalonate,1,3-diethyl 2-isopropylpropanedioate,2-isopropyl-malonic acid diethyl ester
12966
diethyl 2-propan-2-ylpropanedioate
CCOC(=O)C(C(C)C)C(=O)OCC
Infrared Spectrum Authentic
Quantity 25mL
Refractive Index 1.4190 to 1.4210 (20°C, 589nm)
Color Colorless
Packaging Glass bottle
Density 0.981g/mL
Assay Percent Range 98%
Linear Formula (CH3)2CHCH(COOCH2CH3)2
Formula Weight 202.25
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material Diethyl isopropyl malonate
Show More Show Less