Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl methylphosphonate, 96%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass Ampule with Screw-cap
Glass bottle
683-08-9
C5H13O3P
152.13
MFCD00009813
NYYLZXREFNYPKB-UHFFFAOYSA-N
diethyl methylphosphonate,diethyl methanephosphonate,phosphonic acid, methyl-, diethyl ester,demp,1-ethoxy methyl phosphoryl oxyethane,methyl-phosphonicacidiethylester,diethylmethylphosphonate,diethyl methyphosphonate,diethoxymethylphosphine oxide,acmc-1b13h
12685
1-[ethoxy(methyl)phosphoryl]oxyethane
CCOP(=O)(C)OCC
683-08-9
C5H13O3P
152.13
MFCD00009813
NYYLZXREFNYPKB-UHFFFAOYSA-N
diethyl methylphosphonate,diethyl methanephosphonate,phosphonic acid, methyl-, diethyl ester,demp,1-ethoxy methyl phosphoryl oxyethane,methyl-phosphonicacidiethylester,diethylmethylphosphonate,diethyl methyphosphonate,diethoxymethylphosphine oxide,acmc-1b13h
12685
1-[ethoxy(methyl)phosphoryl]oxyethane
CCOP(=O)(C)OCC
For C-C Bond Formation (Olefination)
Packaging Glass Ampule with Screw-cap
Quantity 1g
Refractive Index 1.4130 to 1.4150 (20°C, 589nm)
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96%
Formula Weight 152.13
Physical Form Liquid
Percent Purity ≥95%
Chemical Name or Material Diethyl methylphosphonate, 96%

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 211-667-4

RTECSNumber : SZ9085000