Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Diethyl n-propylmalonate, 99%

Click to view available options
Quantity:
25g
100g
2163-48-6
C10H18O4
202.25
MFCD00009168
GRRSDGHTSMJICM-UHFFFAOYSA-N
diethyl propylmalonate,diethyl 2-propylmalonate,1,3-diethyl 2-propylpropanedioate,diethyl n-propylmalonate,diethylpropylmalonate,propylmalonic acid diethyl ester,propanedioic acid, propyl-, diethyl ester,diethyl-n-propylmalonate,malonic acid, propyl-, diethyl ester,diethyl propyl malonate
16552
diethyl 2-propylpropanedioate
CCCC(C(=O)OCC)C(=O)OCC
2163-48-6
C10H18O4
202.25
MFCD00009168
GRRSDGHTSMJICM-UHFFFAOYSA-N
diethyl propylmalonate,diethyl 2-propylmalonate,1,3-diethyl 2-propylpropanedioate,diethyl n-propylmalonate,diethylpropylmalonate,propylmalonic acid diethyl ester,propanedioic acid, propyl-, diethyl ester,diethyl-n-propylmalonate,malonic acid, propyl-, diethyl ester,diethyl propyl malonate
16552
diethyl 2-propylpropanedioate
CCCC(C(=O)OCC)C(=O)OCC
Applications
Diethyl n-propylmalonate was used in the synthesis of propylacrylic acid (PAA).

Solubility
Soluble in Dichloromethane, Ethanol and Ethyl Acetate.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
Flash Point 91°C (197°F)
Boiling Point 221°C to 222°C
Quantity 25g
Formula Weight 202.25
Percent Purity 99%
Chemical Name or Material Diethyl n-propylmalonate

GHS H Statement
H227
Combustible liquid.

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning