Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Diethyl phenyl orthoformate, 97%

Click to view available options
Quantity:
10g
50g
250g
14444-77-0
C11H16O3
196.246
MFCD00009228
QTURWMMVIIBRRP-UHFFFAOYSA-N
diethyl phenyl orthoformate,diethoxymethoxy benzene,benzene, diethoxymethoxy,diethoxymethoxy-benzene,acmc-209csr,diethoxy phenoxy methane,diethoxymethoxy benzene #,diethylphenyl orthoformate,orthoformic acid diethyl phenyl ester
84440
diethoxymethoxybenzene
CCOC(OCC)OC1=CC=CC=C1
14444-77-0
C11H16O3
196.246
MFCD00009228
QTURWMMVIIBRRP-UHFFFAOYSA-N
diethyl phenyl orthoformate,diethoxymethoxy benzene,benzene, diethoxymethoxy,diethoxymethoxy-benzene,acmc-209csr,diethoxy phenoxy methane,diethoxymethoxy benzene #,diethylphenyl orthoformate,orthoformic acid diethyl phenyl ester
84440
diethoxymethoxybenzene
CCOC(OCC)OC1=CC=CC=C1
Quantity 50g
Flash Point 65°C (149°F)
Boiling Point 103°C to 104°C (10mmHg)
Melting Point -26°C
Beilstein 1872634
Formula Weight 196.25
Percent Purity 97%
Chemical Name or Material Diethyl phenyl orthoformate

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning