Alfa Aesar™ Diethyl phenylphosphonite, 98%

Manufacturer: Alfa Aesar™ B2483614

Quantity 25g

 View more versions of this product

Catalog No. AAB2483614

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1638-86-4
    Molecular Formula (CH3CH2O)2P(C6H5)
    Formula Weight 198.2
    MDL Number MFCD00009086
    Beilstein 2804900
    Synonym Phenylphosphonous acid diethyl ester
    Chemical Name or Material Diethyl phenylphosphonite

    Specifications

    Formula Weight 198.2
    Flash Point >110°C (230°F)
    Quantity 25g
    TSCA TSCA
    Molecular Formula (CH3CH2O)2P(C6H5)
    MDL Number MFCD00009086
    CAS 1638-86-4
    Boiling Point 64°C (0.7mmHg)
    Beilstein 2804900
    Synonym Phenylphosphonous acid diethyl ester
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Recommended Storage 2° to 8°C
    Chemical Name or Material Diethyl phenylphosphonite
    Assay 98%

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    2° to 8°C

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit