Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Diethyl phenylphosphonite, 98%

Click to view available options
Quantity:
5g
25g
100g
1638-86-4
C10H15O2P
198.202
MFCD00009086
RVDJLKVICMLVJQ-UHFFFAOYSA-N
diethyl phenylphosphonite,diethoxyphenylphosphine,phosphonous acid, phenyl-, diethyl ester,diethyl benzenephosphonite,diethyl phenyl-phosphonite,phosphonous acid, p-phenyl-, diethyl ester,pubchem6466,diethylphenylphosphonite,diethyl phenylphophonite,acmc-20al8o
74225
diethoxy(phenyl)phosphane
CCOP(C1=CC=CC=C1)OCC
1638-86-4
C10H15O2P
198.202
MFCD00009086
RVDJLKVICMLVJQ-UHFFFAOYSA-N
diethyl phenylphosphonite,diethoxyphenylphosphine,phosphonous acid, phenyl-, diethyl ester,diethyl benzenephosphonite,diethyl phenyl-phosphonite,phosphonous acid, p-phenyl-, diethyl ester,pubchem6466,diethylphenylphosphonite,diethyl phenylphophonite,acmc-20al8o
74225
diethoxy(phenyl)phosphane
CCOP(C1=CC=CC=C1)OCC
Quantity 100g
Flash Point >110°C (230°F)
Boiling Point 64°C (0.7mmHg)
Beilstein 2804900
Formula Weight 198.2
Percent Purity 98%
Chemical Name or Material Diethyl phenylphosphonite

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Recommended Storage : 2° to 8°C