Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl phosphate, 75%, Tech., balance phosphoric acid, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
598-02-7,7664-38-2
C4H11O4P
154.102
MFCD00039889
UCQFCFPECQILOL-UHFFFAOYSA-N
diethyl phosphate,diethyl acid phosphate,phosphoric acid, diethyl ester,diethyl phosphoric acid,diethylphosphate,diethylphosphoric acid,ethyl phosphate, di,o,o-diethyl hydrogen phosphate,ethyl phosphate, eto 2 ho po,o,o-diethylphosphoric acid
654
CHEBI:27708
diethyl hydrogen phosphate
CCOP(=O)(O)OCC
598-02-7,7664-38-2
C4H11O4P
154.102
MFCD00039889
UCQFCFPECQILOL-UHFFFAOYSA-N
diethyl phosphate,diethyl acid phosphate,phosphoric acid, diethyl ester,diethyl phosphoric acid,diethylphosphate,diethylphosphoric acid,ethyl phosphate, di,o,o-diethyl hydrogen phosphate,ethyl phosphate, eto 2 ho po,o,o-diethylphosphoric acid
654
CHEBI:27708
diethyl hydrogen phosphate
CCOP(=O)(O)OCC
Flash Point 91°C
Color Colorless to Orange-Yellow
Infrared Spectrum Authentic
Melting Point 6.0°C
NMR 70% min.
Boiling Point 203.0°C
Quantity 5g
Density 1.2900g/mL
Packaging Glass bottle
Assay Percent Range 75%
Linear Formula (C2H5O)2PO2H
Formula Weight 154.1
Physical Form Liquid
Percent Purity 75%
Grade Technical
Chemical Name or Material Diethyl phosphate, tech., 75%
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger