Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl propylmalonate, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
Packaging:
Glass bottle
2163-48-6
C10H18O4
202.25
MFCD00009168
GRRSDGHTSMJICM-UHFFFAOYSA-N
diethyl propylmalonate,diethyl 2-propylmalonate,1,3-diethyl 2-propylpropanedioate,diethyl n-propylmalonate,diethylpropylmalonate,propylmalonic acid diethyl ester,propanedioic acid, propyl-, diethyl ester,diethyl-n-propylmalonate,malonic acid, propyl-, diethyl ester,diethyl propyl malonate
16552
diethyl 2-propylpropanedioate
CCCC(C(=O)OCC)C(=O)OCC
2163-48-6
C10H18O4
202.25
MFCD00009168
GRRSDGHTSMJICM-UHFFFAOYSA-N
diethyl propylmalonate,diethyl 2-propylmalonate,1,3-diethyl 2-propylpropanedioate,diethyl n-propylmalonate,diethylpropylmalonate,propylmalonic acid diethyl ester,propanedioic acid, propyl-, diethyl ester,diethyl-n-propylmalonate,malonic acid, propyl-, diethyl ester,diethyl propyl malonate
16552
diethyl 2-propylpropanedioate
CCCC(C(=O)OCC)C(=O)OCC
Packaging Glass bottle
Infrared Spectrum Authentic
Color Colorless
Quantity 5g
Density 0.9800g/mL
Refractive Index 1.4180 to 1.4200 (20°C, 589nm)
Assay Percent Range 99%
Linear Formula C2H5O2CCH(CH2CH2CH3)CO2C2H5
Beilstein 02, 657
Formula Weight 202.25
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material Diethyl propylmalonate, 99%
Show More Show Less

EINECSNumber : 218-492-2