Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl Pyrocarbonate, >97%, Ultrapure, Thermo Scientific Chemicals

Catalog Number AAJ14710AC
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Plastic bottle
1609-47-8
C6H10O5
162.141
MFCD00009106
FFYPMLJYZAEMQB-UHFFFAOYSA-N
diethyl pyrocarbonate, diethyl dicarbonate, depc, diethyl oxydiformate, baycovin, diethylpyrocarbonate, ethyl pyrocarbonate, dicarbonic acid, diethyl ester, dicarbonic acid diethyl ester, diethyl pyrocarbonic acid
3051
CHEBI:59051
ethoxycarbonyl ethyl carbonate
CCOC(=O)OC(=O)OCC
1609-47-8
C6H10O5
162.141
MFCD00009106
FFYPMLJYZAEMQB-UHFFFAOYSA-N
diethyl pyrocarbonate, diethyl dicarbonate, depc, diethyl oxydiformate, baycovin, diethylpyrocarbonate, ethyl pyrocarbonate, dicarbonic acid, diethyl ester, dicarbonic acid diethyl ester, diethyl pyrocarbonic acid
3051
CHEBI:59051
ethoxycarbonyl ethyl carbonate
CCOC(=O)OC(=O)OCC

  • A strong, nonspecific inhibitor of ribonuclease
  • Reacts with the N1 of histidine residues in proteins under mild conditions
  • Used to cross-link and modify proteins by ethoxyformylation of NH2 and imidazole groups in proteins
  • Reacts with amines and thus cannot be used with Tris Buffer

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Strong, nonspecific inhibitor of ribonuclease
  • Reacts with N1 of histidine residues in proteins under mild conditions
  • Used to cross-link and modify proteins by ethoxyformylation of NH2 and imidazole groups in proteins
  • Reacts
TRUSTED_SUSTAINABILITY
Melting Point 69°C
Density 1.101
Boiling Point 93°C to 94°C (18 mmHg)
Flash Point 69°C
Packaging Plastic bottle
Refractive Index 1.398
Quantity 25 mL
Beilstein 637031
Formula Weight 162.14
Percent Purity ≥97%
Grade Ultrapure
Physical Form Liquid
Chemical Name or Material Diethyl Pyrocarbonate
Show More Show Less
Hazard Category H227-H302-H315-H319-H332-H335
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
EINECSNumber 216-542-8
RTECSNumber LQ9350000
TSCA Yes
Recommended Storage Store at -20°C

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit