Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethylaluminium chloride, 0.9M solution in toluene, AcroSeal™, Thermo Scientific Chemicals

Catalog Number p-3212607
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
96-10-6, 108-88-3
C4H10AlCl
120.56
MFCD00000459
YNLAOSYQHBDIKW-UHFFFAOYSA-M
diethylaluminum chloride, aluminum, chlorodiethyl, diethylaluminium chloride, deac, chlorodiethylaluminum, diethylchloroaluminum, diethylaluminum monochloride, deak, aluminum diethyl monochloride, aluminum, dichlorotetraethyldi
7277
chloro(diethyl)alumane
CC[Al](Cl)CC
96-10-6, 108-88-3
C4H10AlCl
120.56
MFCD00000459
YNLAOSYQHBDIKW-UHFFFAOYSA-M
diethylaluminum chloride, aluminum, chlorodiethyl, diethylaluminium chloride, deac, chlorodiethylaluminum, diethylchloroaluminum, diethylaluminum monochloride, deak, aluminum diethyl monochloride, aluminum, dichlorotetraethyldi
7277
chloro(diethyl)alumane
CC[Al](Cl)CC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY
Color Colorless
Density 0.8760g/mL
Flash Point 4°C
Packaging AcroSeal™ Glass Bottle
Linear Formula ClAl(CH2CH3)2
Quantity 100 mL
Specific Gravity 0.876
Solubility Information Solubility in water: reacts
Formula Weight 120.56
Physical Form Liquid
Chemical Name or Material Diethylaluminium chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
May cause damage to organs through prolonged or repeated exposure.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
Suspected of damag
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 202-477-2

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit