Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethylaluminium iodide, 1.1M solution in toluene, AcroSeal™, ACROS Organics™

Click to view available options
Quantity:
800mL
Packaging:
AcroSeal™ Glass Bottle
2040-00-8,108-88-3
C4H10AlI
212.01
MFCD00001025
PPQUYYAZSOKTQD-UHFFFAOYSA-M
aluminum, diethyliodo,diethylaluminum iodide,diethylaluminium iodide,diethyl iodo alumane,et2ali
74874
diethyl(iodo)alumane
CC[Al](CC)I
2040-00-8,108-88-3
C4H10AlI
212.01
MFCD00001025
PPQUYYAZSOKTQD-UHFFFAOYSA-M
aluminum, diethyliodo,diethylaluminum iodide,diethylaluminium iodide,diethyl iodo alumane,et2ali
74874
diethyl(iodo)alumane
CC[Al](CC)I
Lewis acid
Aluminum (Al) 12.2 to 12.7 % (neat product)
Linear Formula (C2H5)2AlI
Density 0.9730g/mL
Formula Weight 212.01
Physical Form Powder or Crystals
Percent Purity 24 to 26 wt%
Iodide 59 to 60 % (neat product)
Infrared Spectrum Authentic
Packaging AcroSeal™ Glass Bottle
Solubility Solubility in water: reacts.
Specific Gravity 0.973
Color Beige to Purple
Quantity 800mL
Flash Point 4°C
Chemical Name or Material Diethylaluminium iodide, 25 wt.% Solution in Toluene
Show More Show Less

GHS H Statement
May be fatal if swallowed and enters airways. Suspected of damaging fertility or the unborn child. May cause drowsiness or dizziness. May cause damage to organs through prolonged or repeated exposure. Causes severe skin burns and eye damage. Highly flammable liquid and vapor. Catches fire spontaneously if exposed to air. Reacts violently with water.

GHS P Statement
Use personal protective equipment as required. Avoid breathing dust/fume/gas/mist/vapors/spray. Do not breathe dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. IF ON SKIN: Wash with plenty of soap and water.

Danger