Alfa Aesar™ Diethylaluminum chloride, 96%, 25% w/w in heptane

Manufacturer: Alfa Aesar™ 89035NF

Quantity 100g

 View more versions of this product

Catalog No. AA89035NF

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 96-10-6
    Molecular Formula C4H10AlCl
    Linear Formula (C2H5)2AlCl
    Formula Weight 120.56
    MDL Number MFCD00000459
    UN Number UN3394
    Chemical Name or Material Diethylaluminum chloride

    Specifications

    Formula Weight 120.56
    Health Hazard 2 GHS H Statement
    H224-H250-H260-H252-H314-H318-H336
    Extremely flammable liquid and vapor.
    Catches fire spontaneously if exposed to air.
    In contact with water releases flammable gases which may ignite spontaneously.
    Self-heating in large quantities
    may catch fire.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    May cause drowsiness or dizziness.
    Health Hazard 3 GHS P Statement
    P210-P303+P361+P353-P305+P351+P338-P405-P422a-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Store contents under inert gas.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C4H10AlCl
    Concentration 25% w/w in heptane
    Physical Form Liquid
    MDL Number MFCD00000459
    Quantity 100g
    Flash Point −18°C (−1°F)
    RTECS Number BD0558000
    TSCA TSCA
    Color Colorless
    Odor Petroleum-like
    DOT Information Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, PYROPHORIC, WATER-REACTIVE
    Sensitivity Air & Moisture Sensitive
    Linear Formula (C2H5)2AlCl
    CAS 96-10-6
    UN Number UN3394
    Health Hazard 1 Danger
    Chemical Name or Material Diethylaluminum chloride
    Assay 96%

    GHS H Statement
    H224-H250-H260-H252-H314-H318-H336
    Extremely flammable liquid and vapor.
    Catches fire spontaneously if exposed to air.
    In contact with water releases flammable gases which may ignite spontaneously.
    Self-heating in large quantities
    may catch fire.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    May cause drowsiness or dizziness.

    GHS P Statement
    P210-P303+P361+P353-P305+P351+P338-P405-P422a-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Store contents under inert gas.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit