Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Diethylaluminum chloride, 96%, 25% w/w in heptane

Manufacturer:  Alfa Aesar™ 89035NB

Quantity 5g

 View more versions of this product

Catalog No. AA89035NB

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

96-10-6
(C2H5)2AlCl
MFCD00000459
Diethylaluminum chloride
C4H10AlCl
120.56
UN3394

Specifications

25% w/w in heptane
5g
Colorless
Air & Moisture Sensitive
C4H10AlCl
120.56
UN3394
96%
96-10-6
−18°C (−1°F)
Petroleum-like
Liquid
(C2H5)2AlCl
MFCD00000459
Diethylaluminum chloride

Safety and Handling

SDS & Safety Documents

GHS H Statement
H224-H250-H260-H252-H314-H318-H336
Extremely flammable liquid and vapor.
Catches fire spontaneously if exposed to air.
In contact with water releases flammable gases which may ignite spontaneously.
Self-heating in large quantities
may catch fire.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause drowsiness or dizziness.

GHS P Statement
P210-P303+P361+P353-P305+P351+P338-P405-P422a-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Store contents under inert gas.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, PYROPHORIC, WATER-REACTIVE

RTECSNumber : BD0558000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit