Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethylamine, 99+%, Thermo Scientific Chemicals

Catalog Number aaa11716ae
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
109-89-7
C4H11N
73.14
MFCD00009032
HPNMFZURTQLUMO-UHFFFAOYSA-N
diethylamine, n,n-diethylamine, ethanamine, n-ethyl, diethamine, diaethylamin, dwuetyloamina, dietilamina, diethylamin, diethyl amine, n-ethyl-ethanamine
8021
CHEBI:85259
CCNCC
109-89-7
C4H11N
73.14
MFCD00009032
HPNMFZURTQLUMO-UHFFFAOYSA-N
diethylamine, n,n-diethylamine, ethanamine, n-ethyl, diethamine, diaethylamin, dwuetyloamina, dietilamina, diethylamin, diethyl amine, n-ethyl-ethanamine
8021
CHEBI:85259
CCNCC

Diethylamine is used as a corrosion inhibitor. It is also used in the production of rubber, resins, dyes and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethylamine is used as a corrosion inhibitor. It is also used in the production of rubber, resins, dyes and pharmaceuticals. It is also used to cook lysergic acid diethylamide (LSD) and is strictly watched by Drug Enforcement Administration (DEA).

Solubility
Miscible with water, alcohol, ether, chloroform and carbon terachloride.

Notes
Air sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point °C to 50°C
Density 0.706
Boiling Point 54°C to 55°C
Flash Point -28°C (-18°F)
Assay Percent Range ≥99%
Refractive Index 1.385
Quantity 100 mL
Beilstein 605268
Sensitivity Air Sensitive
Solubility Information Miscible with water,alcohol,ether,chloroform and carbon terachloride.
IUPAC Name N-ethylethanamine
Formula Weight 73.14
Physical Form Liquid
Chemical Name or Material Diethylamine
Show More Show Less
Hazard Category H225-H302+H332-H311-H314-H335
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: II
EINECSNumber 203-716-3
RTECSNumber HZ8750000
TSCA Yes
storageNote1 Air Sensitive
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit