Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethylamine, 99+%, extra pure, ACROS Organics™

Click to view available options
Quantity:
250mL
1L
2.5L
10L
Packaging:
Glass bottle
Plastic drum
109-89-7
C4H11N
73.13
HPNMFZURTQLUMO-UHFFFAOYSA-N
diethylamine,n,n-diethylamine,ethanamine, n-ethyl,diethamine,diaethylamin,dwuetyloamina,dietilamina,diethylamin,diethyl amine,n-ethyl-ethanamine
8021
CHEBI:85259
N-ethylethanamine
CCNCC
109-89-7
C4H11N
73.13
HPNMFZURTQLUMO-UHFFFAOYSA-N
diethylamine,n,n-diethylamine,ethanamine, n-ethyl,diethamine,diaethylamin,dwuetyloamina,dietilamina,diethylamin,diethyl amine,n-ethyl-ethanamine
8021
CHEBI:85259
N-ethylethanamine
CCNCC
Viscosity 0.34 mPa.s (25°C)
Quantity 250mL
Packaging Glass bottle
Infrared Spectrum Authentic
Density 0.7100g/mL
Water 0.1% Max. (K.F.)
Melting Point -50.0°C
Boiling Point 55.0°C
Flash Point −23°C
Color Colorless
Specific Gravity 0.71
Refractive Index 1.3840 to 1.3860
Assay Percent Range 99+%
Linear Formula (C2H5)2NH
Beilstein 04, 95
Fieser 04,148; 13,312; 14,191
Merck Index 15, 3122
Solubility Information Solubility in water: soluble. Other solubilities: miscible with alcohol and most common organic, solvents
Formula Weight 73.13
Physical Form Liquid
Percent Purity ≥99%
Grade Extra Pure
Chemical Name or Material Diethylamine
Show More Show Less

GHS P Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Highly flammable liquid and vapor.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN: Wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger