Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethylamine, Reagent, 98%, Spectrum™

Click to view available options
Quantity:
500 mL
2.5 L
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly-Coated Glass Bottle
Steel Can
109-89-7
C4H11N
73.14
HPNMFZURTQLUMO-UHFFFAOYSA-N
diethylamine
CCNCC
109-89-7
C4H11N
73.14
HPNMFZURTQLUMO-UHFFFAOYSA-N
diethylamine
CCNCC

DI117, 109-89-7, C4H11N

Spectrum™ Diethylamine, Reagent is a secondary amine that is a flammable, volatile, strongly alkaline, colorless liquid that has a strong unpleasant smell. It is used in the production of rubber, resin, dyes and pharmaceuticals and as a corrosion inhibiter. Its Reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.
Packaging Steel Can
Quantity 20 L
CAS Max % 1
UN Number UN1154
Percent Purity 98%