Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Diethylaminosulfur trifluoride, 95%

Manufacturer:  Alfa Aesar™ A1199206

Quantity 5g

 View more versions of this product

Catalog No. AAA1199206


  Check Availability
Add to cart

Specifications

Description

Applications
Diethylaminosulfur trifluoride (DAST) is used as a precursor to prepare 2,6-dimethoxybenzoyl fluoride and 2-thiazolines. It is a useful fluorinating agent used for a variety of compounds including alcohols, thioethers, alkenols and cyanohydrins. It also serves as a catalyst for Friedel-Crafts allylation reaction using tertiary cyclopropyl silyl ethers and in the rearrangement of homoallylic alcohols to unsaturated aldehydes. It plays an important role for gem difluorination of ketopipecolinic acids.

Solubility
Miscible with water, ethanol, benzene and acetonitrile.

Notes
Store in cool place. Moisture sensitive. It reacts with water and ethanol. Incompatible with water, acids and bases.
Description & Specifications

Chemical Identifiers

38078-09-0
(CH3CH2)2NSF3
MFCD00000363
1849066
Diethylaminosulfur trifluoride
C4H10F3NS
161.19
UN2920
DAST

Specifications

5g
C4H10F3NS
161.19
UN2920
1849066
DAST
95%
38078-09-0
(CH3CH2)2NSF3
MFCD00000363
30° to 32°C (3mmHg)
23°C (73°F)
Diethylaminosulfur trifluoride

Safety and Handling

SDS & Safety Documents

GHS H Statement
H331-H314-H318-H226
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.

GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310-P402
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: I; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.

Recommended Storage : 2°C to 8°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit