Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Diethylaminosulfur trifluoride, 95%, ACROS Organics™
Click to view available options
Quantity:
5g
25g
100g
Packaging:
Plastic bottle
Description
C-X bond formation reagent
Specifications
Specific Gravity | 1.316 |
Packaging | Plastic bottle |
Refractive Index | 1.4100 to 1.4160 |
NMR | 94% min. |
Quantity | 25g |
Flash Point | 23°C |
Boiling Point | 30.0°C to 32.0°C (3.0mmHg) |
Infrared Spectrum | Authentic |
Density | 1.3160g/mL |
Color | Brown to Yellow |
Show More |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Flammable liquid and vapour.
Harmful in contact with skin.
Harmful if inhaled.
Reacts violently with water.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open f
GHS Signal Word: Danger
EINECSNumber : 253-771-2