Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Diethyldithiocarbamic acid, sodium salt trihydrate, ACS reagent, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 95.0°C to 98.5°C |
Color | Gray to White |
Infrared Spectrum | Authentic |
Packaging | Glass bottle |
Linear Formula | (C2H5)2NCS2Na·3H2O |
Quantity | 100 g |
Beilstein | 04, II, 613 |
Sensitivity | (to copper) Passes Test |
Merck Index | 15, 3421 |
Solubility Information | (in water) Passes test |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Warning |
Hazard Statement | GHS H Statement Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. Very toxic to aquatic life. |
Precautionary Statement | GHS P Statement Do not eat,drink or smoke when using this product. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if presen |
RTECSNumber | EZ6550000 |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction