Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl bis(hydroxymethyl)malonate, 95%, Acros Organics

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
20605-01-0
C9H16O6
220.22
MFCD00009130
WIOHBOKEUIHYIC-UHFFFAOYSA-N
diethyl bis hydroxymethyl malonate,diethyl 2,2-bis hydroxymethyl malonate,propanedioic acid, bis hydroxymethyl-, diethyl ester,1,3-diethyl 2,2-bis hydroxymethyl propanedioate,diethyl 2,2-bis hydroxymethyl propanedioate,2,2-bis hydroxymethyl malonic acid diethyl ester,bis hydroxymethyl malonic acid diethyl ester,diethyl 2,2-bis hydroxymethyl propane-1,3-dioate,acmc-20al8x,ksc490a8h
311844
diethyl 2,2-bis(hydroxymethyl)propanedioate
CCOC(=O)C(CO)(CO)C(=O)OCC
20605-01-0
C9H16O6
220.22
MFCD00009130
WIOHBOKEUIHYIC-UHFFFAOYSA-N
diethyl bis hydroxymethyl malonate,diethyl 2,2-bis hydroxymethyl malonate,propanedioic acid, bis hydroxymethyl-, diethyl ester,1,3-diethyl 2,2-bis hydroxymethyl propanedioate,diethyl 2,2-bis hydroxymethyl propanedioate,2,2-bis hydroxymethyl malonic acid diethyl ester,bis hydroxymethyl malonic acid diethyl ester,diethyl 2,2-bis hydroxymethyl propane-1,3-dioate,acmc-20al8x,ksc490a8h
311844
diethyl 2,2-bis(hydroxymethyl)propanedioate
CCOC(=O)C(CO)(CO)C(=O)OCC
Flash Point >110°C
Quantity 100g
Color White
Melting Point 45°C to 52°C
Packaging Glass bottle
Infrared Spectrum Authentic
Assay Percent Range 95%
Linear Formula (HOCH2)2C(CO2C2H5)2
Formula Weight 220.22
Physical Form Crystalline Solid
Percent Purity 95%
Chemical Name or Material Diethyl bis(hydroxymethyl)malonate
Show More Show Less

GHS H Statement
Causes skin irritation.

GHS Signal Word: Warning

EINECSNumber : 243-916-8