Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl isocyanomethylphosphonate, 97%, Acros Organics

Click to view available options
Quantity:
1mL
5mL
Packaging:
Glass bottle
41003-94-5
C6H12NO3P
177.14
MFCD00042951
ICURVXBGGDVHDT-UHFFFAOYSA-N
diethyl isocyanomethylphosphonate,diethylisocyanomethylphosphonate,acmc-20al8q,diethoxycarbonyl methanisocyanide,diethyl isocyanomethyl phosphonate,diethoxyphosphinylmethyl isocyanide,isocyanomethylphosphonic acid diethyl ester,isocyanomethylphosphononic acid diethyl ester,1-ethoxy isocyanomethyl phosphoryl oxyethane
2734699
1-[ethoxy(isocyanomethyl)phosphoryl]oxyethane
CCOP(=O)(C[N+]#[C-])OCC
41003-94-5
C6H12NO3P
177.14
MFCD00042951
ICURVXBGGDVHDT-UHFFFAOYSA-N
diethyl isocyanomethylphosphonate,diethylisocyanomethylphosphonate,acmc-20al8q,diethoxycarbonyl methanisocyanide,diethyl isocyanomethyl phosphonate,diethoxyphosphinylmethyl isocyanide,isocyanomethylphosphonic acid diethyl ester,isocyanomethylphosphononic acid diethyl ester,1-ethoxy isocyanomethyl phosphoryl oxyethane
2734699
1-[ethoxy(isocyanomethyl)phosphoryl]oxyethane
CCOP(=O)(C[N+]#[C-])OCC
for C-C Bond Formation (Olefination)
Boiling Point 84°C to 87°C (0.1mmHg)
Flash Point >110°C
Specific Gravity 1.105
Density 1.1050g/mL
Refractive Index 1.4310 to 1.4340 (20°C, 589nm)
Packaging Glass bottle
Color Colorless to Yellow
Infrared Spectrum Authentic
Quantity 5mL
Assay Percent Range 96% min. (GC)
Linear Formula CNCH2P(O)(OC2H5)2
Fieser 14,135
Formula Weight 177.14
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Diethyl isocyanomethylphosphonate
Show More Show Less

GHS H Statement
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a

GHS Signal Word: Warning