Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Diethylene glycol dibutyl ether

Catalog No. 2056211L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-562-11L 1 L
20-562-1500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 20-562-11L Supplier Sigma Aldrich Supplier No. 2056211L
Add to Cart
Add to Cart
This item is not returnable. View return policy

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Diethylene glycol dibutyl ether

Specifications

CAS 112-73-2
Melting Point -60°C (lit.)
Density 0.885 g/mL (at 25°C (literature))
Boiling Point 256°C (lit.)
Molecular Formula C12H26O3
Refractive Index n20/D 1.423 (literature)
Linear Formula (CH3CH2CH2CH2OCH2CH2)2 O
MDL Number MFCD00009459
Quantity 1 L
Synonym 2-Butoxyethyl ether; Bis(2-butoxyethyl) ether; Dibutyldiglycol
Molecular Weight (g/mol) 218.33
Percent Purity ≥99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.