Alfa Aesar™ Diethylene glycol dibutyl ether, 99+%

Extraction of precious metals

Manufacturer: Alfa Aesar™ 42150AC

Quantity 25mL

 View more versions of this product

Catalog No. AA42150AC

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 112-73-2
    Formula Weight 218.74
    MDL Number MFCD00009459
    Synonym Dibutyl carbitol; Butyl diglyme
    Chemical Name or Material Diethylene glycol dibutyl ether

    Specifications

    Formula Weight 218.74
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00009459
    Synonym Dibutyl carbitol; Butyl diglyme
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Refractive Index 1.4235
    Physical Form Liquid
    Boiling Point 256°C
    Flash Point 117° (243°F)
    Density 0.885g/mL
    Quantity 25mL
    Color Colorless
    CAS 112-73-2
    Melting Point −60°C
    Health Hazard 1 Warning
    TSCA TSCA
    RTECS Number KN0350000
    Chemical Name or Material Diethylene glycol dibutyl ether
    Assay ≥99%

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit