Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethylene glycol dibutyl ether, 99+%, pure, ACROS Organics™

Click to view available options
Quantity:
250mL
1L
2.5L
Packaging:
Glass bottle
112-73-2
C12H26O3
218.34
KZVBBTZJMSWGTK-UHFFFAOYSA-N
diethylene glycol dibutyl ether,bis 2-butoxyethyl ether,dibutyl carbitol,butyl diglyme,1-2-2-butoxyethoxy ethoxy butane,5,8,11-trioxapentadecane,ether, bis butoxyethyl,2,2'-dibutoxyethyl ether,ether, bis 2-butoxyethyl
8210
1-[2-(2-butoxyethoxy)ethoxy]butane
CCCCOCCOCCOCCCC
112-73-2
C12H26O3
218.34
KZVBBTZJMSWGTK-UHFFFAOYSA-N
diethylene glycol dibutyl ether,bis 2-butoxyethyl ether,dibutyl carbitol,butyl diglyme,1-2-2-butoxyethoxy ethoxy butane,5,8,11-trioxapentadecane,ether, bis butoxyethyl,2,2'-dibutoxyethyl ether,ether, bis 2-butoxyethyl
8210
1-[2-(2-butoxyethoxy)ethoxy]butane
CCCCOCCOCCOCCCC
Viscosity 2.39 mPa.s (20°C)
Quantity 1L
Melting Point -60.0°C
Color Undesignated
Boiling Point 256.0°C
Density 0.8800g/mL
Refractive Index 1.4225 to 1.4245
Water 0.1% Max. (K.F.)
Specific Gravity 0.88
Infrared Spectrum Authentic
Flash Point 118°C
Packaging Glass bottle
Assay Percent Range 99% min. (GC)
Linear Formula [CH3(CH2)3OCH2CH2]2O
Solubility Information Solubility in water: moderate soluble (3g/L at 20°C). Other solubilities: soluble in chloroform
Formula Weight 218.34
Physical Form Liquid
Percent Purity 99+%
Grade Pure
Chemical Name or Material Diethylene glycol dibutyl ether
Show More Show Less

EINECSNumber : 204-001-9