Alfa Aesar™ Diethylenetriaminepentaacetic acid, 98+%

Manufacturer: Alfa Aesar™ A10926-18

Quantity 50g

 View more versions of this product

Catalog No. AAA1092618

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 67-43-6
    Molecular Formula C14H23N3O10
    Formula Weight 393.35
    MDL Number MFCD00004289
    Beilstein 1810219
    Merck Index 14,7125
    Synonym DTPA; Pentetic acid
    Chemical Name or Material Diethylenetriaminepentaacetic acid

    Specifications

    Formula Weight 393.35
    CAS 67-43-6
    Merck Index 14,7125
    Synonym DTPA; Pentetic acid
    Odor Odorless
    Molecular Formula C14H23N3O10
    Melting Point ≈220°C (dec.)
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for sev
    TSCA TSCA
    Beilstein 1810219
    Quantity 50g
    MDL Number MFCD00004289
    RTECS Number MB8205000
    Chemical Name or Material Diethylenetriaminepentaacetic acid
    Assay ≥98%

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for sev

    Warning

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit