Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethylenetriaminepentaacetic Acid, Pentasodium Salt, 41% Solution in Water, Spectrum™

Click to view available options
Quantity:
100 mL
500 mL
4 L
Packaging:
Poly Bottle
140-01-2
C14H18N3Na5O10
503.26
LQPLDXQVILYOOL-UHFFFAOYSA-I
pentasodium 2-[bis({2-[bis(carboxylatomethyl)amino]ethyl})amino]acetate
[Na+].[Na+].[Na+].[Na+].[Na+].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CCN(CC([O-])=O)CC([O-])=O
140-01-2
C14H18N3Na5O10
503.26
LQPLDXQVILYOOL-UHFFFAOYSA-I
pentasodium 2-[bis({2-[bis(carboxylatomethyl)amino]ethyl})amino]acetate
[Na+].[Na+].[Na+].[Na+].[Na+].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CCN(CC([O-])=O)CC([O-])=O

D1156, 140-01-2, C14H18N3Na5O10

Spectrum™ Diethylenetriaminepentaacetic Acid, Pentasodium Salt, 41 Percent Solution in Water is also known under the abbreviation DETA and is a colorless liquid that is hygroscopic and soluble in both water and organic solvent that are polarized. Used as a tridentate ligand forming complex in coordination chemistry, it is also used for the extraction of gases that are acid in the oil industry.
Packaging Poly Bottle
Quantity 4 L
CAS Max % 1
Percent Purity 39 to 41%