Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl phenylmalonate, 98%, ACROS Organics™

Click to view available options
Quantity:
5mL
100mL
250mL
1L
2.5L
Packaging:
Glass bottle
83-13-6
C13H16O4
236.267
MFCD00009144
FGYDHYCFHBSNPE-UHFFFAOYSA-N
diethyl phenylmalonate,diethyl 2-phenylmalonate,phenylmalonic acid diethyl ester,propanedioic acid, phenyl-, diethyl ester,diethylphenylmalonate,1,3-diethyl 2-phenylpropanedioate,malonic acid, phenyl-, diethyl ester,propanedioic acid, 2-phenyl-, 1,3-diethyl ester,diethyl 2-phenylpropane-1,3-dioate,diethyl-phenylmalonate
66514
diethyl 2-phenylpropanedioate
CCOC(=O)C(C1=CC=CC=C1)C(=O)OCC
83-13-6
C13H16O4
236.267
MFCD00009144
FGYDHYCFHBSNPE-UHFFFAOYSA-N
diethyl phenylmalonate,diethyl 2-phenylmalonate,phenylmalonic acid diethyl ester,propanedioic acid, phenyl-, diethyl ester,diethylphenylmalonate,1,3-diethyl 2-phenylpropanedioate,malonic acid, phenyl-, diethyl ester,propanedioic acid, 2-phenyl-, 1,3-diethyl ester,diethyl 2-phenylpropane-1,3-dioate,diethyl-phenylmalonate
66514
diethyl 2-phenylpropanedioate
CCOC(=O)C(C1=CC=CC=C1)C(=O)OCC
Water 0.05% max.
Refractive Index 1.4903 to 1.4923
Color Colorless to Yellow
Infrared Spectrum Authentic
Packaging Glass bottle
Boiling Point 170.0°C to 172.0°C (14.0mmHg)
Density 1.0900g/mL
Quantity 1L
Melting Point 16.0°C to 17.0°C
Flash Point >112°C
Assay Percent Range 98%
Linear Formula C2H5O2CCH(C6H5)CO2C2H5
Beilstein 09, 854
Formula Weight 236.27
Physical Form Liquid After Melting
Percent Purity 98%
Chemical Name or Material Diethyl phenylmalonate
Show More Show Less