Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl phenylmalonate, 98%, Thermo Scientific Chemicals

Catalog Number AC150262500
Click to view available options
Quantity:
5 mL
100 mL
250 mL
1 L
2.5 L
Packaging:
Glass bottle
83-13-6
MFCD00009144
FGYDHYCFHBSNPE-UHFFFAOYSA-N
diethyl phenylmalonate, diethyl 2-phenylmalonate, phenylmalonic acid diethyl ester, propanedioic acid, phenyl-, diethyl ester, diethylphenylmalonate, 1,3-diethyl 2-phenylpropanedioate, malonic acid, phenyl-, diethyl ester, propanedioic acid, 2-phenyl-, 1,3-diethyl ester, diethyl 2-phenylpropane-1,3-dioate, diethyl-phenylmalonate
66514
diethyl 2-phenylpropanedioate
CCOC(=O)C(C1=CC=CC=C1)C(=O)OCC
83-13-6
MFCD00009144
FGYDHYCFHBSNPE-UHFFFAOYSA-N
diethyl phenylmalonate, diethyl 2-phenylmalonate, phenylmalonic acid diethyl ester, propanedioic acid, phenyl-, diethyl ester, diethylphenylmalonate, 1,3-diethyl 2-phenylpropanedioate, malonic acid, phenyl-, diethyl ester, propanedioic acid, 2-phenyl-, 1,3-diethyl ester, diethyl 2-phenylpropane-1,3-dioate, diethyl-phenylmalonate
66514
diethyl 2-phenylpropanedioate
CCOC(=O)C(C1=CC=CC=C1)C(=O)OCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 16.0°C to 17.0°C
Color Colorless to Yellow
Density 1.0900g/mL
Boiling Point 170.0°C to 172.0°C (14.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C13H16O4
Refractive Index 1.4903 to 1.4923
Linear Formula C2H5O2CCH(C6H5)CO2C2H5
Quantity 250 mL
Beilstein 09, 854
Specific Gravity 1.09
Solubility Information Solubility in water: immiscible
Molecular Weight (g/mol) 236.27
Formula Weight 236.27
Percent Purity 98%
Physical Form Liquid After Melting
Chemical Name or Material Diethyl phenylmalonate
Show More Show Less
EINECSNumber 201-456-5

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit