Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DiethyleVinylphosphonate, 97%, ACROS Organics™

Click to view available options
Quantity:
10g
25g
100g
Packaging:
Glass bottle
682-30-4
C6H13O3P
164.14
MFCD00009079
DREPONDJUKIQLX-UHFFFAOYSA-N
diethyl vinylphosphonate,vinylphosphonic acid diethyl ester,diethyl vinylphosphate,diethoxyphosphinylethene,phosphonic acid, vinyl-, diethyl ester,diethyl ethenylphosphonate,phosphonic acid, ethenyl-, diethyl ester,1-ethenyl ethoxy phosphoryl oxyethane,acmc-1at5g
69629
1-[ethenyl(ethoxy)phosphoryl]oxyethane
CCOP(=O)(C=C)OCC
682-30-4
C6H13O3P
164.14
MFCD00009079
DREPONDJUKIQLX-UHFFFAOYSA-N
diethyl vinylphosphonate,vinylphosphonic acid diethyl ester,diethyl vinylphosphate,diethoxyphosphinylethene,phosphonic acid, vinyl-, diethyl ester,diethyl ethenylphosphonate,phosphonic acid, ethenyl-, diethyl ester,1-ethenyl ethoxy phosphoryl oxyethane,acmc-1at5g
69629
1-[ethenyl(ethoxy)phosphoryl]oxyethane
CCOP(=O)(C=C)OCC
for C-C Bond Formation (Olefination)
Density 1.0600g/mL
Refractive Index 1.4280 to 1.43
Boiling Point 202°C
Specific Gravity 1.06
Flash Point >110°C
Packaging Glass bottle
Quantity 100g
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Linear Formula (C2H5O)2P(O)CH=CH2
Formula Weight 164.14
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material Diethyl vinylphosphonate
Show More Show Less