Alfa Aesar™ Diethylgermanium dichloride

As an intermediate

Manufacturer: Alfa Aesar™ 3410803

Quantity 1g

 View more versions of this product

Catalog No. AA3410803

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 13314-52-8
    Formula Weight 201.64
    MDL Number MFCD00013589
    UN Number UN3265
    Chemical Name or Material Diethylgermanium dichloride

    Specifications

    Formula Weight 201.64
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Solubility Information Decomposes in water and NH3; Soluble in organic solvents
    MDL Number MFCD00013589
    CAS 13314-52-8
    UN Number UN3265
    Density 1.372g/mL
    Color Colorless
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
    Boiling Point 175°C
    Melting Point −37°C
    Physical Form Liquid
    Health Hazard 1 Danger
    Refractive Index 1.4710
    Quantity 1g
    Health Hazard 2 GHS H Statement
    H314-H318-H302-H312-H332-H227
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Combustible liquid.
    Odor Acrid
    Sensitivity Moisture Sensitive
    Flash Point 63° (145°F)
    Chemical Name or Material Diethylgermanium dichloride

    GHS H Statement
    H314-H318-H302-H312-H332-H227
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Combustible liquid.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit