Alfa Aesar™ Diethylmethylvinylsilane, 98+%

Manufacturer: Alfa Aesar™ B2321414

Quantity 25g

 View more versions of this product

Catalog No. AAB2321414

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 18292-29-0
    Molecular Formula C7H16Si
    Linear Formula (CH3CH2)2CH3SiCH=CH2
    Formula Weight 128.29
    MDL Number MFCD00041344
    UN Number UN1993
    Synonym Vinyldiethylmethylsilane
    Chemical Name or Material Diethylmethylvinylsilane

    Specifications

    Formula Weight 128.29
    Molecular Formula C7H16Si
    Linear Formula (CH3CH2)2CH3SiCH=CH2
    MDL Number MFCD00041344
    CAS 18292-29-0
    UN Number UN1993
    Synonym Vinyldiethylmethylsilane
    Boiling Point 118° to 119°C
    Quantity 25g
    Flash Point 1°C (35°F)
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H225
    Highly flammable liquid and vapour.
    Health Hazard 3 GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    Chemical Name or Material Diethylmethylvinylsilane
    Assay ≥98%

    GHS H Statement
    H225
    Highly flammable liquid and vapour.

    GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit