Alfa Aesar™ Diethylphenylphosphine, 97%

Manufacturer: Alfa Aesar™ 1018006

Quantity 5g

 View more versions of this product

Catalog No. AA1018006

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1605-53-4
    Formula Weight 166.21
    MDL Number MFCD00015172
    UN Number UN3278
    Chemical Name or Material Diethylphenylphosphine

    Specifications

    Formula Weight 166.21
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P304+P340-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H331-H227-H302-H312
    Toxic if inhaled.
    Combustible liquid.
    Harmful if swallowed.
    Harmful in contact with skin.
    Physical Form Liquid
    MDL Number MFCD00015172
    Health Hazard 1 Danger
    Refractive Index 1.5475
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOPHOSPHORUS COMPOUND, LIQUID, TOXIC, N.O.S.
    Quantity 5g
    UN Number UN3278
    Density 0.954g/mL
    Flash Point 66° (150°F)
    Boiling Point 220°C
    CAS 1605-53-4
    Odor Pungent
    Sensitivity Air & Moisture Sensitive
    Chemical Name or Material Diethylphenylphosphine
    Assay 97%

    GHS H Statement
    H331-H227-H302-H312
    Toxic if inhaled.
    Combustible liquid.
    Harmful if swallowed.
    Harmful in contact with skin.

    GHS P Statement
    P210-P261-P280-P304+P340-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit