Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Diethylphenylphosphine, 97%

Manufacturer:  Alfa Aesar™ 1018014

Quantity 25g

 View more versions of this product

Catalog No. AA1018014


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

1605-53-4
MFCD00015172
Diethylphenylphosphine
166.21
UN3278

Specifications

0.954g/mL
Pungent
220°C
1605-53-4
Liquid
MFCD00015172
Diethylphenylphosphine
66° (150°F)
Air & Moisture Sensitive
25g
1.5475
166.21
UN3278
97%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H331-H227-H302-H312
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P210-P261-P280-P304+P340-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOPHOSPHORUS COMPOUND, LIQUID, TOXIC, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit