Alfa Aesar™ Diethylvinylphosphonate, 97%

Manufacturer: Alfa Aesar™ 1036804

Quantity 2g

 View more versions of this product

Catalog No. AA1036804

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 682-30-4
    Formula Weight 164.14
    MDL Number MFCD00009079
    UN Number UN3278
    Chemical Name or Material Diethylvinylphosphonate

    Specifications

    Formula Weight 164.14
    Health Hazard 3 GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Liquid
    UN Number UN3278
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOPHOSPHORUS COMPOUND, LIQUID, TOXIC, N.O.S.
    Boiling Point 202°C
    MDL Number MFCD00009079
    CAS 682-30-4
    Refractive Index 1.4290
    Density 1.068g/mL
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301
    Toxic if swallowed.
    Flash Point 110° (230°F)
    Quantity 2g
    Chemical Name or Material Diethylvinylphosphonate
    Assay 97%

    GHS H Statement
    H301
    Toxic if swallowed.

    GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit