Learn More
Diethylzinc, 1.5M solution in toluene, AcroSeal™, Thermo Scientific Chemicals
$187.70 - $618.50
Specifications
Chemical Name or Material | Diethylzinc |
---|---|
Color | Colorless |
Name Note | 1.5M Solution in Toluene |
CAS | 557-20-0, 108-88-3 |
Molecular Formula | C4H10Zn |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
AC377311000
|
Thermo Scientific Chemicals
377311000 |
100 mL | AcroSeal™ Glass bottle |
Each for $187.70
Order in Bulk
Request bulk or custom formats |
|
||||
AC377318000
|
Thermo Scientific Chemicals
377318000 |
800 mL | AcroSeal™ Glass Bottle |
Each for $618.50
Order in Bulk
Request bulk or custom formats |
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Diethylzinc | |
1.5M Solution in Toluene | |
C4H10Zn | |
MFCD00009021 | |
0.9130g/mL | |
diethylzinc, zinc, diethyl, diethyl zinc, zinc ethide, zinc ethyl, unii-s0w5nqh7c6, et2zn, znet2, s0w5nqh7c6, c2h5 2zn | |
HQWPLXHWEZZGKY-UHFFFAOYSA-N | |
diethylzinc | |
11185 | |
0.913 |
Colorless | |
557-20-0, 108-88-3 | |
(C2H5)2Zn | |
1.4 to 1.6M | |
15, 3151 | |
Solubility in water: reacts violently. Other solubilities: miscible with most hydrocarbons | |
CC[Zn]CC | |
123.50 | |
123.5 | |
Liquid |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor. In contact with water releases flammable gases which may ignite spontaneously. May be fatal if swallowed and enters airways. Causes severe skin burns and eye damage. May cause drowsiness or dizziness. Suspected of damaging the unborn child. May cause damage to organs through prolonged or repeated exposure. Toxic to aquatic life with long lasting effects. Reacts violently with water.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Store in a dry place. Store in a closed container.
Danger
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.