Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Diethylzinc, nominally 15% w/w in hexanes, Thermo Scientific™
Click to view available options
Quantity:
100 g
500 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Color | Brown to Colorless |
Density | 0.720 g/mL |
Flash Point | −40°C (−40°F) |
Linear Formula | Zn(C2H5)2 |
Quantity | 500 g |
UN Number | UN3394 |
Sensitivity | Air and moisture sensitive |
Solubility Information | Reacts with water. |
IUPAC Name | zinc;ethane |
Formula Weight | 123.49 |
Show More |
Safety and Handling
Hazard Category | H225-H250-H260-H304-H314-H335-H336-H361f-H373 |
Hazard Statement | GHS H Statement H225-H250-H260-H252-H304-H361-H373-H314-H336 Highly flammable liquid and vapor. Catches fire spontaneously if exposed to air. In contact with water releases flammable gases which may ignite spontaneously. Self-heating in large quantities may catch fire. May be fatal if swallowed and enters airways. Suspected of damaging fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Causes severe skin burns and eye damage. May cause drowsiness or dizziness. |
Precautionary Statement | P201-P202-P210-P222-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P281-P302+P334-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, PYROPHORIC, WATER-REACTIVE |
EINECSNumber | 209-161-3 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Argon |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction