Learn More
Medchemexpress LLC Difenoconazole (Standard) | 119446-68-3 | 95.6% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB0850
Encompass_Preferred
Difenoconazole (Standard) is the analytical standard of Difenoconazole. This product is intended for research and analytical applications. Difenoconazole is a sterol demethylation inhibitor, acting as a fungicide. It binds to the heme portion of the fungal cytochrome P450 51, which interferes with mycelial growth and inhibits the spore germination of pathogens, thereby suppressing fungal growth.
  • Analytical standard of difenoconazole.
  • Intended for research and analytical applications.
  • Sterol demethylation inhibitor.
  • Acts as a fungicide.
  • Interferes with mycelial growth.
  • Inhibits the spore germination of pathogens.
  • Suppresses fungal growth.
  • Used in qualitative, quantitative, and methodological research experiments.
  • Applied in techniques such as HPLC, GC, and MS.
Catalog No. 50-004-50498