Learn More
Andwin ScientificSupplier Diversity Partner DIFF QUIK STAIN KIT DFS Item
Manufacturer:  Andwin ScientificSupplier Diversity Partner B41321A
The Diff-quik Stain Set is composed of 500mL each of diff-quik fixative and Solutions 1 and 2, which are both ideal for use for hematology and cytology stain and histological applications.
Catalog No. NC9943455
  Check Availability
Add to cart
For SAVINGS of $658.91, you may also be interested in considering Fisher HealthCare™ PROTOCOL™ Hema 3™ Manual Staining System and Stat Pack Available on GSA/VA Contract for Federal Government customers only. as an alternative option to catalog number NC9943455
Show Details
22-122911
Hema 3 System; 3 x 500mL