Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diflunisal, USP, 98-101.5%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Poly Bottle
Poly Pail
22494-42-4
C13H8F2O3
250.20
HUPFGZXOMWLGNK-UHFFFAOYSA-N
2',4'-difluoro-4-hydroxy-[1,1'-biphenyl]-3-carboxylic acid
OC(=O)C1=CC(=CC=C1O)C1=CC=C(F)C=C1F
22494-42-4
C13H8F2O3
250.20
HUPFGZXOMWLGNK-UHFFFAOYSA-N
2',4'-difluoro-4-hydroxy-[1,1'-biphenyl]-3-carboxylic acid
OC(=O)C1=CC(=CC=C1O)C1=CC=C(F)C=C1F

D1160, 22494-42-4, C13H8F2O3

Spectrum™ Diflunisal, USP is classified as a non-steroidal anti-inflammatory drug. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Poly Bottle
Ignition Residue 0.001
Loss on Drying 0.003
Quantity 100 g
CAS Max % 1
Percent Purity 98 to 101.5%