Alfa Aesar™ Difurfuryl sulfide, 98%

Manufacturer: Alfa Aesar™ B2199914

Quantity 25g

 View more versions of this product

Catalog No. AAB2199914

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 13678-67-6
    Molecular Formula C10H10O2S
    Formula Weight 194.25
    MDL Number MFCD00010081
    Beilstein 1371700
    Synonym Furfuryl sulfide; 2,2'-(Thiodimethylene)difuran
    Chemical Name or Material Difurfuryl sulfide

    Specifications

    Formula Weight 194.25
    TSCA TSCA
    Molecular Formula C10H10O2S
    MDL Number MFCD00010081
    CAS 13678-67-6
    Beilstein 1371700
    Synonym Furfuryl sulfide; 2,2'-(Thiodimethylene)difuran
    Flash Point >110°C (230°F)
    Odor Stench
    Boiling Point 139° to 140°C (14mmHg)
    Melting Point 30° to 32°C
    Quantity 25g
    Chemical Name or Material Difurfuryl sulfide
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit