Learn More
Digi-Sense™ Borescopes
Cole Parmer Digi-Sense Borescopes features a flexible, camera-tipped probe. Ideal for inspecting duct work, wiring locations, piping and vehicles.
 View more versions of this product
Catalog No. 13-245-282
Add to cart
Includes: Probe, Mirror, Hook, Magnet, Carrying case, USB cable, DC 5V charger, Video output cable, Rechargable 3.7V lithium battery
Description
Description
  • Camera-tipped probe provides easy real-time viewing on color LCD monitor in hard-to-reach areas
  • Bright LCD monitor allows instant viewing of area to be inspected
  • 180° image rotation on LCD
  • Adjustable LED light on camera tip provides the necessary light in dimly lit areas or adjusts down when viewing reflective objects to eliminate washout
Compliance
CE; IP67
Warranty and Services
2 year
Specifications
Specifications
Built-in lithium battery
8GB flash memory
English, French, German, Spanish
TV-out hookup
3.2in.
Yes
CE; IP67
Color LCD monitor
4 adjustable LED lights
Battery; USB charging
Still and video images for playback