Learn More
Digi-Sense™ Borescopes
Cole Parmer Digi-Sense Borescopes features a flexible, camera-tipped probe. Ideal for inspecting duct work, wiring locations, piping and vehicles.
$218.43
Includes: Probe, Mirror, Hook, Magnet, Carrying case, USB cable, DC 5V charger, Video output cable, Rechargable 3.7V lithium battery
Products 2
Catalog Number Data Storage Video Output Price Quantity & Availability  
Catalog Number Data Storage Video Output Price Quantity & Availability  
13-245-282
View Documents
8GB flash memory Still and video images for playback Each for $218.43
Add to cart
13-245-283
View Documents
No No This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Part Not Found
N/A
Description
Description
  • Camera-tipped probe provides easy viewing in hard-to-reach areas
  • Bright LCD monitor allows instant viewing of area to be inspected
  • Adjustable LED light on camera tip provides the necessary light in dimly lit areas or adjusts down when viewing reflective objects to eliminate washout
Compliance
CE; IP67
Warranty and Services
2 year