Learn More
Digi-Sense™ Precalibrated Professional Thermocouple Thermometers
Rugged, heavy-duty construction protects against knocks and jars
 View more versions of this product
Catalog No. 13-245-276
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Includes: Type K bead wire temperature probe (one for each input), hard carrying case, one 9 V battery, and NIST-traceable calibration report with data supplied by InnoCal. Data logging models 20250-19 and -20 also include USB cable and software.
Description
Description
Reliable measurements, guaranteed! Precalibrated to NIST-traceable standards|Relative time stamp provides a time reference for major events|Electronic off set function allows compensation of probe errors|Infrared input port allows safe, noncontact measurements with optional IR wand
Warranty and Services
2 Years
Specifications
Specifications
-328°F to +2501°F
2500°F
−328°F
Precalibrated
8 in.
Type K bead wire temperature probe (one for each input), hard carrying case, one 9 V battery, and NIST-traceable calibration report with data supplied by InnoCal. 
0.1° 1° above 1000°
None
Backlit LCD
-200°C to +1372°C
1372°C
−200°C
Any Type-K thermocouple probes with mini connector
20.32 cm
Type K Thermocouple
2-input Thermocouple Thermometer
±0.15% of reading +1.8°F; ±0.5% of reading +3.6°F below -148°F [±0.15% of reading +1.0°C; ±0.5% of reading +2.0°C below -100°C]