Learn More
Motic Swift Line Digital 1.2 MP Microscope Camera Products for Science Education
Easy enough for the microscopy novice, but sophisticated enough for the professional. With back illuminated sensor technology and speedy data transfer, this camera creates superbly detailed images and smooth video captures.
Manufacturer:  Motic Swift Line DMOTICAM S1
Catalog No. S81793
Add to cart
Description
Description
Motic's new C-mount ready sCMOS cameras are the newest members of the Moticam family of professionally designed and engineered cameras. Easy enough for the microscopy novice, but sophisticated enough for the professional. With back illuminated sensor technology and speedy data transfer, this camera creates superbly detailed images and smooth video captures.
Included Motic Images software enables live and captured images at a maximum quality. Edit, measure, and share images or reports with this comprehensive software.
INCLUDES
  • CS ring
  • Mini USB 3.1 cable (removable)
  • Motic 4-dot Calibration slide
  • Motic Images Plus 3.0 for PC and Mac
CAMERA SPECIFICATIONS
  • 1.2 MP 1/3 in. sCMOS
  • USB 3.1
  • 1280 x 960 @ 120 fps
  • 640 x 480 @ 240 fps
REQUIRED SPECIFICATIONS
  • Processor: 2 Ghz multi-core or above
  • USB 2.0 (Minimum Requirement)
  • USB 3.0 (Required for high speed transmission)
  • RAM: 8 GB
  • Hard Disk: 5 GB (Free Space)
  • Video Card: MUST have dedicated 1 GB Video RAM
Specifications
Specifications
1.2 MP
USB Microscope Camera